Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Citric Acid Anhydrous, Purified, Reagents

Catalog No. 01337600
Change view
Click to view available options
Quantity:
500 g
1 kg
3 kg
10 kg
50 kg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-337-600 50 kg
01-337-604 10 kg
01-337-603 1 kg
01-337-602 3 kg
01-337-601 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 01-337-600 Supplier Reagents Holdings Llc Supplier No. C210455050F4
Add to Cart
Add to Cart

Citric Acid Anhydrous, Purified, Chemically Pure, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 77-92-9
Molecular Formula C6H8O7
Molecular Weight (g/mol) 192.12
InChI Key KRKNYBCHXYNGOX-UHFFFAOYSA-N
Synonym 2-hydroxypropane-1,2,3-tricarboxylic Acid
IUPAC Name 2-hydroxypropane-1,2,3-tricarboxylic acid
SMILES OC(=O)CC(O)(CC(O)=O)C(O)=O

Specifications

Melting Point 153°C
Boiling Point 175°C
Color Colorless/White
Grade Pure
Quantity 50 kg
Packaging Fiber Drum
Physical Form Solid
Specific Gravity 1.665
Chemical Name or Material Citric Acid Anhydrous, Purified
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.