Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Citric acid monohydrate, 99+%, ACS reagent

Catalog No. p-4380078
Click to view available options
Quantity:
250 g
1 kg
2.5 kg
Packaging:
Plastic bottle

Catalog No. AC385852500

Supplier: Thermo Scientific Chemicals 385852500

Only null left
Add to Cart

CAS: 5949-29-1 | C6H8O7·H2O | 210.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5949-29-1
Molecular Formula C6H8O7·H2O
Molecular Weight (g/mol) 210.15
InChI Key YASYEJJMZJALEJ-UHFFFAOYSA-N
Synonym citric acid monohydrate, citric acid hydrate, 2-hydroxypropane-1,2,3-tricarboxylic acid hydrate, citric acid, monohydrate, unii-2968phw8qp, 1,2,3-propanetricarboxylic acid, 2-hydroxy-, monohydrate, citrate, acidum citricum monohydricum, citric acid monohydrate usp
PubChem CID 22230
ChEBI CHEBI:31404
IUPAC Name 2-hydroxypropane-1,2,3-tricarboxylic acid;hydrate
SMILES C(C(=O)O)C(CC(=O)O)(C(=O)O)O.O

Specifications

Melting Point 135.0°C to 152.0°C
Flash Point 174°C
Ignition Residue 0.02% max
Assay Percent Range 99+%
Packaging Plastic bottle
Linear Formula HO2CCH2C(OH)(CO2H)CH2CO2H·H2O
Quantity 250 g
Merck Index 15,2325
Solubility Information Solubility in water: 676g/L (25°C). Other solubilities: 2.17g/100g ether - 0.007g/100g chloroform, 15.43g/100g amyl alcohol - 5.98g/100g amyl acetate, 5.28g/100g ethyl acetate - 197g/100g methanol, 62.8g/100g propanol
Formula Weight 210.15
Percent Purity 99+%
Grade ACS Reagent
Insoluble Matter 0.005% max
Chemical Name or Material Citric acid monohydrate
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes serious eye irritation.
Precautionary Statement GHS P Statement
Wash face,hands and any exposed skin thoroughly after handling.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact len

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.