Learn More
Citric Acid, Monohydrate, USP, EP, BP, JP, FCC, bioCERTIFIED™, 12 kg, Spectrum Chemical
Click to view available options
:
Quantity: 12 kg
Description
Citric Acid, Monohydrate, USP, EP, BP, JP, FCC, bioCERTIFIED &rade; is used as an acidulant to control pH and to passivate high purity process piping. All Spectrum Chemical USP, EP, BP, JP, FCC bioCERTIFIED™ grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP).
Specifications
Specifications
Quantity | 12 kg |