Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(-)-β-Citronellol, ≥98.5% (sum of enantiomers, GC), MilliporeSigma™ Supelco™

Catalog No. p-7187712 Shop All MilliporeSigma Supelco Products
Click to view available options
Quantity:
1 mL
5 mL

Catalog No. 11-101-5436

Supplier: MilliporeSigma Supelco 274831MLF

Only null left
Add to Cart
Add to Cart

Citronellol is a monoterpene alcohol found in essential oils of Cymbopogon winterianus Jowitt. S-(−)-isomer that is less common. It is prevalent in geranium and citronella oils.

Featured Industries: Cleaning Products, Cosmetics, Food and Beverages and Personal Care
TRUSTED_SUSTAINABILITY

Specifications

CAS 7540-51-4
Density 0.854 g/mL (at 20°C)
Flash Point 99°C - Closed Cup
Physical Form Neat
Linear Formula C10H20 O
MDL Number MFCD00063214
Boiling Point 221°C to 224°C
Synonym beta-Rhodinol; (S)-(-)-beta-Citronellol; (S)-3,7-Dimethyl-6-octen-1-ol
Type Analytical Standard
Quantity 1 mL
Formula Weight 156.27
Percent Purity ≥98.5% (sum of enantiomers, GC)
Grade Analytical Standard
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.