Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

citrullinated RGG Rabbit anti-Human, Unconjugated, Polyclonal, MilliporeSigma™

Catalog No. ABS224410MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ABS224410MI 100 μg
ABS224425MI 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ABS224410MI Supplier MilliporeSigma Supplier No. ABS2244100UG
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

citrullinated RGG Polyclonal antibody specifically detects citrullinated RGG in Human samples. It is validated for ELISA, Western Blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen citrullinated RGG
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Gene Alias Deaminated RGG
Host Species Rabbit
Immunogen A chemically synthesized citrullinated peptide (CGGCitGGGCitGG).
Molecular Weight of Antigen Multiple bands based on multiple protein citrullination. Uncharacterized bands may be observed in some lysate(s).
Purification Method Affinity Purified
Quantity 100 μg
Research Discipline Inflammation and Immunology
Primary or Secondary Primary
Test Specificity This rabbit polyclonal antibody specifically detects citrullinated RGG motifs in proteins.
Target Species Human
Content And Storage Stable for 1 year at 2 to 8°C from date of receipt.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.