Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Citrulline Monoclonal Antibody (2D3.1), FITC
GREENER_CHOICE

Catalog No. PIMA545754
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA545754 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIMA545754 Supplier Invitrogen™ Supplier No. MA545754
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

A 1:1,000 dilution of MA5-45754 was sufficient for detection of Citrulline in 2 μg of Citrulline conjugated to BSA by ECL immunoblot analysis using Goat Anti-Mouse IgG:HRP as the secondary Antibody. Specific for citrulline residues on modified proteins. Tested using citrulline conjugated Bovine Serum Albumin (BSA).

Citrulline is an amino acid that is not incorporated into proteins during synthesis but is generated within proteins by the deimination of arginine. Citrulline occurs normally in many proteins, such as Myelin basic protein (MBP) while others, such as fibrin become citrullinated during inflammation or apoptosis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Citrulline
Applications ELISA, Immunohistochemistry, Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 2D3.1
Concentration 1 mg/mL
Conjugate FITC
Formulation PBS, 9.09mM sodium bicarbonate with 640.91mM DMSO, 136.36mM ethanolamine and no preservative; pH 7.4
Gene Alias ACPA; Citrulline; L-Citrulline
Host Species Mouse
Immunogen Synthetic L-Citrulline conjugated to Keyhole Limpet Hemocyanin (KLH).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Chemical
Content And Storage 4°C, store in dark
Product Type Antibody
Form Liquid
Isotype IgG1
Target Molecule Chemical
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.