Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Citrulline Mouse, PerCP, Clone: 2D3.1, Abnova™

Catalog No. 89338711 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-338-711 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-338-711 Supplier Abnova Corporation Supplier No. MAB19207
Add to Cart
Add to Cart

Mouse Monoclonal Antibody.

Citrulline Monoclonal antibody specifically detects Citrulline in samples. It is validated for WB, ELISA, ICC, IF.

Specifications

Antigen Citrulline
Applications Western Blot, ELISA, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone 2D3.1
Conjugate PerCP
Host Species Mouse
Immunogen Synthetic L-Citrulline conjugated to Keyhole Limpet Hemocyanin (KLH)
Quantity 100 μg
Primary or Secondary Primary
Content And Storage Store at -20°C
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.