Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CIZ Polyclonal Antibody, Bethyl Laboratories

Catalog No. 501564656
Encompass_Preferred
Change view
Click to view available options
Quantity:
20 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-156-4656 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-156-4656 Supplier Bethyl Laboratories, Inc Supplier No. A303251AT
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Rat

This gene contains long CAG trinucleotide repeats coding consecutive glutamine residues. The gene product may functions as a transcription factor, with a potential role in the regulation of neurodevelopment or neuroplasticity. The protein appears to bind and regulate the promoters of MMP1, MMP3, MMP7 and COL1A1. Studies in mouse suggest that nuclear matrix transcription factors (NP/NMP4) may be part of a general mechanical pathway that couples cell construction and function during extracellular matrix remodeling. Multiple transcript variants encoding several isoforms have been found for this gene.

Specifications

Antigen CIZ
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation TBS with 0.1% BSA and 0.09% sodium azide
Gene ZNF384
Gene Accession No. Q8TF68
Gene Alias CAGH1, CAGH1A, CIZ, ERDA2, NMP4, NP, TNRC1, CAG repeat protein 1, Cas-interacting zinc finger protein, expanded repeat domain, CAG/CTG 2, nuclear matrix transcription factor 4, trinucleotide repeat-containing gene 1 protein
Gene Symbols ZNF384
Host Species Rabbit
Immunogen Between 527 and 577
Purification Method Antigen affinity chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 171017
Target Species Human
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.