Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CIZ1 Antibody, Alexa Fluor™ 647, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB10074624AF647
Description
CIZ1 Polyclonal antibody specifically detects CIZ1 in Human, Mouse samples. It is validated for Western Blot, ImmunofluorescenceSpecifications
CIZ1 | |
Polyclonal | |
Western Blot, Immunocytochemistry/Immunofluorescence | |
CDKN1A interacting zinc finger protein 1, CDKN1A-interacting zinc finger protein 1, cip1-interacting zinc finger protein, LSFR1ZNF356Nuclear protein NP94, NP94, Zinc finger protein 356 | |
The immunogen recognized by this antibody maps to a region between residue 848 and 898 of human CDKN1A interacting zinc finger protein 1 using the numbering given in entry NP_036259.2 (GeneID 25792). | |
0.1 mL | |
Cancer | |
25792 | |
Store at 4C in the dark. | |
IgG |
Western Blot, Immunofluorescence | |
Alexa Fluor 647 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human, Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction