Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CIZ1 Antibody, Novus Biologicals™

Goat Polyclonal Antibody
Supplier: Novus Biologicals NBP226207
Description
CIZ1 Polyclonal specifically detects CIZ1 in Human samples. It is validated for Western Blot, Peptide ELISA.Specifications
CIZ1 | |
Polyclonal | |
Unconjugated | |
Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide | |
CIZ1 | |
Peptide with sequence C-QPNTQDKTPSKVTAR corresponding to C-Terminus according to NP_036259.2, NP_001124487.1, NP_001124489.1, NP_001124490.1, NP_001244904.1, NP_001244905.1. | |
0.1 mg | |
Cancer | |
25792 | |
Human | |
IgG |
Western Blot, ELISA | |
0.5 mg/mL | |
Western Blot 0.3 - 1 ug/ml, Peptide ELISA Detection limit 1:128000 | |
CDKN1A interacting zinc finger protein 1, CDKN1A-interacting zinc finger protein 1, cip1-interacting zinc finger protein, LSFR1ZNF356Nuclear protein NP94, NP94, Zinc finger protein 356 | |
Goat | |
Affinity Purified | |
RUO | |
Primary | |
This antibody is expected to recognize all reported isoforms (NP_036259.2; NP_001124487.1; NP_001124489.1; NP_001124490.1; NP_001244904.1; NP_001244905.1). Reported variants represent identical protein: NP_036259.2, NP_001124488.1 | |
Store at -20C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction