Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CIZ1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA541533
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA541533 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA541533 Supplier Invitrogen™ Supplier No. PA541533
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: YKAAKNPSPT TRPVSRRCAI NARNALTALF TSSGRPPSQP NTQDKTPSKV Sequence homology: Horse: 77%; Human: 100%.

May regulate the subcellular localization of CIP/WAF1.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CIZ1
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene CIZ1
Gene Accession No. Q9ULV3
Gene Alias 0610038H21Rik; 2900056O04Rik; CDKN1A interacting zinc finger protein 1; CDKN1A-interacting zinc finger protein 1; Cip1-interacting zinc finger protein; CIZ1; LSFR1; NP94; nuclear protein NP94; Zinc finger protein 356; ZNF356
Gene Symbols CIZ1
Host Species Rabbit
Immunogen Synthetic peptide directed towards the C-terminal of human CIZ1 (aa 831-880).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 25792
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.