Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CIZ1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5109546
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5109546 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5109546 Supplier Invitrogen™ Supplier No. PA5109546
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PGQLQVKAQP QARMTVPKQT QTPDLLPEAL EAQVLPRFQP RVLQVQAQVQ SQTQPRIPST DTQVQPKLQK QAQTQTSPEH LVLQQKQVQP QLQQEAEPQK QVQPQVHTQA QPSVQPQEHP PAQVSVQPPE QTHEQPHTQP QVSLLAPEQT PVVVHVCGLE MPPDAVEAGG GMEKTLPEPV GTQVSMEEIQ N.

May regulate the subcellular localization of CIP/WAF1.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CIZ1
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.55 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene CIZ1
Gene Accession No. Q9ULV3
Gene Alias 0610038H21Rik; 2900056O04Rik; CDKN1A interacting zinc finger protein 1; CDKN1A-interacting zinc finger protein 1; Cip1-interacting zinc finger protein; CIZ1; LSFR1; NP94; nuclear protein NP94; Zinc finger protein 356; ZNF356
Gene Symbols CIZ1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 250-440 of human CIZ1 (NP_001124490).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 25792, 68379
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.