Learn More
CIZ1 Rabbit anti-Human, Polyclonal, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A301497A

Description
The recommended shelf life for this product is 1 year from date of receipt.
May regulate the subcellular localization of CIP/WAF1.Specifications
CIZ1 | |
Polyclonal | |
Unconjugated | |
CIZ1 | |
LSFR1, NP94, ZNF356, CDKN1A-interacting zinc finger protein 1, cip1-interacting zinc finger protein, nuclear protein NP94, zinc finger protein 356 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
25792 | |
4° C | |
Liquid |
Immunoprecipitation, Western Blot | |
1 mg/ml | |
phosphate, tris citrate with 0.09% sodium azide; pH 7-8 | |
Q9ULV3 | |
CIZ1 | |
Between 848 and 898 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.