Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ CK 666
GREENER_CHOICE

Catalog No. 395050
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
39-505-0 50 mg
39-501-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 39-505-0 Supplier Tocris Bioscience™ Supplier No. 3950/50
Add to Cart
Add to Cart
This item is not returnable. View return policy

Arp2/3 inhibitor; inhibits actin polymerization

Inhibitor of the Arp2/3 complex; inhibits actin polymerization (IC50 = 4 μM).

Chemical Identifiers

CAS 442633-00-3
Molecular Formula C18H17FN2O

Specifications

Quantity 50 mg
Formula Weight 296.34
Purity >99%
Molecular Weight (g/mol) 296.345
Chemical Name or Material CK 666
Recommended Storage Store at -20°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.