Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pointe Scientific CK Reagents

Catalog No. 23666214 Shop All Pointe Scientific Products
Change view
Click to view available options
Quantity:
3 x 150 R2mL
4 x 500 R1mL
Type:
Creatine Kinase R1 Kit
Creatine Kinase R2 Kit
For Use With (Equipment):
Hitachi 747 analyzers
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Type For Use With (Equipment)
23-666-214 4 x 500 R1mL Creatine Kinase R1 Kit Hitachi 747 analyzers
23-666-215 3 x 150 R2mL Creatine Kinase R2 Kit Hitachi 747 analyzers
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 23-666-214 Supplier Pointe Scientific Supplier No. HC422R1

Specifications

Certifications/Compliance CE Marked
Type Creatine Kinase R1 Kit
Formulation CK R1 (Buffer) contains: Imidazole Buffer 100mM/L (pH 6.7), NADP 2mM/L, HK (Baker's Yeast) 2.5KU/L, Glucose 20mM/L, Magnesium Acetate 10mM/L, EDTA 2mM/L, N-acetylcysteine 20mM/L
For Use With (Application) Quantitative determination of Creatine Kinase activity in Serum
For Use With (Equipment) Hitachi 747 analyzers
Quantity 4 x 500 R1mL
Form Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.