Learn More
Promega CK1 alpha 1 Kinase Enzyme System, 10microg
Catalog No. PRV4484 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
10 μg
Catalog No. Quantity
PRV4484 10 μg
1 options
Catalog No. PRV4484 Supplier Promega Supplier No. V4484
Full-length recombinant human CK1 alpha 1 was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. CK1 alpha 1 is a member of the CK1 family of serine/threonine protein kinases, which play an important role in diverse cell processes.
  • Easily Screen and Profile CK1α1 Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP−Glo™ Assay for bioluminescent detection of kinase activity
Specifications
Quantity 10 μg