Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CK1 delta Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5109600
Description
Immunogen sequence: TYLNFCRSLR FDDKPDYSYL RQLFRNLFHR QGFSYDYVFD WNMLKFGASR AADDAERERR DREERLRHSR NPATRGLPST ASGRLRGTQE VAPPTPLTPT S.
CSNK1D (CK1 delta), a serine/threonine kinase, plays a critical role in Wnt/beta-catenin signaling pathway. This pathway is important in both development and cancer. The encoded protein may also be involved in the regulation of apoptosis, circadian rhythm, microtubule dynamics, chromosome segregation, and p53-mediated effects on growth. The encoded protein is highly similar to the mouse and rat CK1 delta homologs. Three transcript variants encoding different isoforms have been found for CK1 delta.
Specifications
| CK1 delta | |
| Polyclonal | |
| Unconjugated | |
| CSNK1D | |
| 1200006A05Rik; AA409348; ASPS; casein kinase 1; casein kinase 1 delta; casein kinase 1, delta; casein kinase I isoform delta; CK1d; CK1delta; CKId; CKIdelta; CKI-delta; CSNK1D; D930010H05Rik; FASPS2; HCKID; HGNC:2452; Tau-protein kinase CSNK1D | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 64462 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 2.28 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| Q06486 | |
| CSNK1D | |
| A synthetic peptide corresponding to a sequence within amino acids 250-350 of human CSNK1D (NP_620693.1). | |
| 100 μL | |
| Primary | |
| Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction