Learn More
Promega CK1 epsilon Kinase Enzyme System, 10microg
Catalog No. PRV4160 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
10 μg
Catalog No. Quantity
PRV4160 10 μg
1 options
Catalog No. PRV4160 Supplier Promega Supplier No. V4160
Full-length recombinant human CK1 epsilon was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. CK1 epsilon is a member of the CK1 family of serine/threonine protein kinases, which play an important role in diverse cell processes.
  • Easily Screen and Profile CK1ε Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP−Glo™ Assay for bioluminescent detection of kinase activity
Specifications
Quantity 10 μg
Content And Storage <-65°C