Learn More
Promega CK1 epsilon Kinase Enzyme System, 1mg
Catalog No. PRV6286 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
1 mg
Catalog No. Quantity
PRV6286 1 mg
1 options
Catalog No. PRV6286 Supplier Promega Supplier No. V6286
Add to Cart
Add to Cart
Request a Quote
Full-length recombinant human CK1 epsilon was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. CK1 epsilon is a member of the CK1 family of serine/threonine protein kinases, which play an important role in diverse cell processes.
  • Easily Screen and Profile CK1ε Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP−Glo™ Assay for bioluminescent detection of kinase activity
Specifications
Quantity 1 mg
Content And Storage <-65°C