Learn More
Invitrogen™ CK1 epsilon Monoclonal Antibody (OTI5C9)

Description
CK1 epsilon Monoclonal Antibody for Western Blot
Specifications
Specifications
| Antigen | CK1 epsilon |
| Applications | Western Blot |
| Classification | Monoclonal |
| Clone | OTI5C9 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | CSNK1E |
| Gene Accession No. | P49674 |
| Gene Alias | AI426939; AI551861; AW457082; casein kinase 1; casein kinase 1 epsilon; casein kinase 1, epsilon; casein kinase I isoform epsilon; CK1e; CK1epsilon; CKI, epsilon; CKIe; CKIepsilon; CKI-epsilon; Csnk1e; HCKIE; KC1 epsilon; KC1epsilon; MGC10398; OTTHUMP00000063258; OTTHUMP00000063263; RP1-5O6.1; tau |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.