Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CK1 epsilon Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA595934
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA595934 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA595934 Supplier Invitrogen™ Supplier No. PA595934
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: EFSTYLNFCR SLRFDDKPDY SYLRQLFRNL FHRQGFSYDY VFDWNMLKFG AARNPEDVDR ERREHEREER MGQLRGSATR ALPPGPPTGA TANRLRSAAE PVASTPASRI QPAGNTSPRA ISRVDRERKV SMRLHRGAPA NVSSSDLTGR QEVSRIPASQ TSVPFDHLGK; Positive Samples: HeLa, 293T; Cellular Location: Cytoplasm, Nucleus.

CSNK1E (CK1 epsilon) is a serine/threonine kinase implicated in a wide range of cellular functions, including vesicular trafficking, DNA damage repair, cell cycle progression, and cytokinesis. CSNK1E phosphorylate key regulatory proteins involved in the control of cell differentiation, proliferation, chromosome segregation, circadian rhythms, and metabolic pathways. CK1 family members share a highly conserved kinase domain but differ in their variable N-and C-terminal domains. Mammals have seven family members: alpha, beta, gamma1, gamma2, gamma3, delta and epsilon. CKIe positively regulates Akt possibly by inhibiting PP2A. CKIe play an important role in neurodegenerative diseases.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CK1 epsilon
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.93 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene CSNK1E
Gene Accession No. P49674, Q9JMK2
Gene Alias AI426939; AI551861; AW457082; casein kinase 1; casein kinase 1 epsilon; casein kinase 1, epsilon; casein kinase I isoform epsilon; CK1e; CK1epsilon; CKI, epsilon; CKIe; CKIepsilon; CKI-epsilon; Csnk1e; HCKIE; KC1 epsilon; KC1epsilon; MGC10398; OTTHUMP00000063258; OTTHUMP00000063263; RP1-5O6.1; tau
Gene Symbols CSNK1E
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 247-416 of human CSNK1E (NP_6894071).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1454, 27373, 58822
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.