Learn More
Description
- Easily Screen and Profile CK1γ1 Kinase Inhibitors
- Includes kinase, substrate and reaction buffer
- Use with ADP−Glo™ Assay for bioluminescent detection of kinase activity
Specifications
Specifications
| Quantity | 10 μg |
| Content And Storage | CK1γ1 Kinase, 10μg (Human, recombinant; amino acids 21 to end). MW: ∽70 - 76kDa; Native Casein Protein Substrate, purified from bovine milk; Reaction Buffer, DTT Store below -65°C |
| For Use With (Equipment) | ADP-Glo™ Assay for bioluminescent detection of kinase activity |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.