Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CK1 gamma-1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA599014
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA599014 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA599014 Supplier Thermo Scientific Supplier No. PA599014
Add to Cart
Add to Cart

CK1 gamma-1 Polyclonal Antibody

CK1 gamma-1 Polyclonal Antibody for Western Blot, IHC (P), ELISA

TRUSTED_SUSTAINABILITY

Specifications

Antigen CK1 gamma-1
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 3.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene Csnk1g1
Gene Accession No. Q9HCP0
Gene Alias 9130020E21Rik; casein kinase 1 gamma 1; casein kinase 1, gamma 1; casein kinase I isoform gamma-1; casein kinase I, gamma 1; CK I gamma1; CK1G1; CK1gamma1; CKI gamma 1; CKI gamma1; CK-I gamma1; CKI- gamma1; CKI gamma-1; CKI-gamma 1; CKI-gamma-1; CSNK1G1; KC1G1
Gene Symbols Csnk1g1
Host Species Rabbit
Immunogen Recombinant Human Casein kinase I isoform gamma-1 protein (1-125AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 53944
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.