Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CK1 gamma-1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5104268
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5104268 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5104268 Supplier Invitrogen™ Supplier No. PA5104268
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total Casein Kinase Igamma1.

Casein kinase I (also designated CKI) and casein kinase II (also designated CKII) compose a family of serine/ threonine protein kinases which are present in all eukaryotes examined to date. CKI family members, which include CKI alpha, gamma, epsilon and delta, have been implicated in the control of cytoplasmic and nuclear processes, including DNA replication and repair. CKII is usually expressed as a tetrameric complex consisting of either an alpha2 beta2 or an alpha alpha' beta2 structure. The alpha catalytic subunit is stimulated by the beta regulatory subunit, which undergoes autophosphorylation. CKII activity is high in the cytosol and nucleus of proliferating and differentiating cells. CKII is known to phosphorylate more than 100 different substrates including nuclear oncoproteins, transcription factors and enzymes involved in DNA metabolism.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CK1 gamma-1
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene Csnk1g1
Gene Accession No. Q62761, Q8BTH8, Q9HCP0
Gene Alias 9130020E21Rik; casein kinase 1 gamma 1; casein kinase 1, gamma 1; casein kinase I isoform gamma-1; casein kinase I, gamma 1; CK I gamma1; CK1G1; CK1gamma1; CKI gamma 1; CKI gamma1; CK-I gamma1; CKI- gamma1; CKI gamma-1; CKI-gamma 1; CKI-gamma-1; CSNK1G1; KC1G1
Gene Symbols Csnk1g1
Host Species Rabbit
Immunogen A synthesized peptide derived from human CSNK1G1(Accession Q9HCP0), corresponding to amino acid residues P336-A386.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 214897, 53944, 64086
Target Species Human, Mouse, Rat, Monkey
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.