Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CK1 gamma-2 Monoclonal Antibody (OTI4H3)
GREENER_CHOICE

Catalog No. PIMA526560
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA526560 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA526560 Supplier Invitrogen™ Supplier No. MA526560
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CK1 gamma-2 Monoclonal Antibody for Western Blot, IHC (P)

Casein kinases are operationally defined by their preferential utilization of acidic proteins such as caseins as substrates. It can phosphorylate a large number of proteins. Participates in Wnt signaling.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CK1 gamma-2
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI4H3
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene Csnk1g2
Gene Accession No. P78368
Gene Alias 2810429I12Rik; AI463719; casein kinase 1 gamma 2; casein kinase 1 gamma 2 isoform; casein kinase 1 isoform gamma-2; casein kinase 1, gamma 2; casein kinase I isoform gamma-2; CK Igamma2; CK1G2; CKI gamma 2; CKI gamma2; CKI-gamma 2; CKIgamma2; CK-Igamma2; CKI-gamma2; CKI-gamma-2; CKIgamma-2; Csnk1g2
Gene Symbols Csnk1g2
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 163-415 of CSNK1G2 produced in E.coli.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1455
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.