Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CK1 gamma-2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA586401
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA586401 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA586401 Supplier Invitrogen™ Supplier No. PA586401
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Purity is > 95% by SDS-PAGE.

Casein kinases are operationally defined by their preferential utilization of acidic proteins such as caseins as substrates. It can phosphorylate a large number of proteins. Participates in Wnt signaling.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CK1 gamma-2
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.2
Gene Csnk1g2
Gene Accession No. P78368, Q62762, Q8BVP5
Gene Alias 2810429I12Rik; AI463719; casein kinase 1 gamma 2; casein kinase 1 gamma 2 isoform; casein kinase 1 isoform gamma-2; casein kinase 1, gamma 2; casein kinase I isoform gamma-2; CK Igamma2; CK1G2; CKI gamma 2; CKI gamma2; CKI-gamma 2; CKIgamma2; CK-Igamma2; CKI-gamma2; CKI-gamma-2; CKIgamma-2; Csnk1g2
Gene Symbols Csnk1g2
Host Species Rabbit
Immunogen Synthetic peptide, corresponding to the N-terminal of Human Casein Kinase Igamma2.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 103236, 1455, 65278
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.