Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CK1 gamma-3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA582950
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA582950 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA582950 Supplier Invitrogen™ Supplier No. PA582950
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MFDYEYDWIG KQLPTPVGAV QQDPALSSNR EAHQHRDKMQ QSKNQVVSST NGELNTDDPT AGRSNAPITA PTEVEVMDET NCQKV Highest antigen sequence indentity to the following orthologs: Mouse - 99%, Rat - 68%.

The casein kinase I (CKI) gene family encodes serine/threonine kinases that preferentially phosphorylate acidic substrates. CKI proteins are monomeric and range from 25 to 55 kD. They are ubiquitous, being found in the nuclei, cytoplasm, and membrane fractions of eukaryotic cells. In eukaryotic cells, CKI is in volved in regulating cellular metabolism, transcription, cell cycle progression, cytoskeletal rearrangement and cell movement, apoptosis, and differentiation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CK1 gamma-3
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene CSNK1G3
Gene Accession No. Q9Y6M4
Gene Alias 3300002K07Rik; C330049O21Rik; casein kinase 1 gamma 3; casein kinase 1, gamma 3; casein kinase I isoform gamma-3; CKI-gamma 3; CSNK1G3; CSNK1G3L
Gene Symbols CSNK1G3
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human CSNK1G3. Recombinant protein control fragment (Product #RP-91455).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1456
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.