Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CK1 gamma-3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA588914
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA588914 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA588914 Supplier Invitrogen™ Supplier No. PA588914
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LRKLFTDLFD RKGYMFDYEY DWIGKQLPTP VGAVQQDPAL SSNREAHQHR DKMQQSKNQV VSSTNGELNT DDPTAGRSNA PITAPTEVEV MDETNCQKVL NMWCCCFFKR RKRKTIQRHK; Positive Samples: Mouse eye, Mouse brain, Rat heart; Cellular Location: Cytoplasm.

The casein kinase I (CKI) gene family encodes serine/threonine kinases that preferentially phosphorylate acidic substrates. CKI proteins are monomeric and range from 25 to 55 kD. They are ubiquitous, being found in the nuclei, cytoplasm, and membrane fractions of eukaryotic cells. In eukaryotic cells, CKI is in volved in regulating cellular metabolism, transcription, cell cycle progression, cytoskeletal rearrangement and cell movement, apoptosis, and differentiation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CK1 gamma-3
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.68 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene CSNK1G3
Gene Accession No. Q62763, Q8C4X2
Gene Alias 3300002K07Rik; C330049O21Rik; casein kinase 1 gamma 3; casein kinase 1, gamma 3; casein kinase I isoform gamma-3; CKI-gamma 3; CSNK1G3; CSNK1G3L
Gene Symbols CSNK1G3
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 304-423 of human CSNK1G3 (NP_0010269821).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 64823, 70425
Target Species Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.