Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CK1 gamma-3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA599838
Description
Antibody detects endogenous levels of total CSNK1G3.
The casein kinase I (CKI) gene family encodes serine/threonine kinases that preferentially phosphorylate acidic substrates. CKI proteins are monomeric and range from 25 to 55 kD. They are ubiquitous, being found in the nuclei, cytoplasm, and membrane fractions of eukaryotic cells. In eukaryotic cells, CKI is in volved in regulating cellular metabolism, transcription, cell cycle progression, cytoskeletal rearrangement and cell movement, apoptosis, and differentiation.
Specifications
CK1 gamma-3 | |
Polyclonal | |
Unconjugated | |
CSNK1G3 | |
3300002K07Rik; C330049O21Rik; casein kinase 1 gamma 3; casein kinase 1, gamma 3; casein kinase I isoform gamma-3; CKI-gamma 3; CSNK1G3; CSNK1G3L | |
Rabbit | |
Affinity chromatography | |
RUO | |
1456, 64823, 70425 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q62763, Q8C4X2, Q9Y6M4 | |
CSNK1G3 | |
A synthesized peptide derived from human CSNK1G3(Accession Q9Y6M4), corresponding to amino acid residues M1-P41. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction