Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CK1 gamma Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA571858
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA571858 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA571858 Supplier Invitrogen™ Supplier No. PA571858
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

CK1 gamma Polyclonal Antibody for Western Blot, IHC (P), Flow

Casein kinase I (also designated CKI) and casein kinase II (also designated CKII) compose a family of serine/ threonine protein kinases which are present in all eukaryotes examined to date. CKI family members, which include CKI alpha, gamma, epsilon and delta, have been implicated in the control of cytoplasmic and nuclear processes, including DNA replication and repair. CKII is usually expressed as a tetrameric complex consisting of either an alpha2 beta2 or an alpha alpha' beta2 structure. The alpha catalytic subunit is stimulated by the beta regulatory subunit, which undergoes autophosphorylation. CKII activity is high in the cytosol and nucleus of proliferating and differentiating cells. CKII is known to phosphorylate more than 100 different substrates including nuclear oncoproteins, transcription factors and enzymes involved in DNA metabolism.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CK1 gamma
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene CSNK1G2
Gene Accession No. Q8BTH8, Q8BVP5, Q8C4X2
Gene Alias 2810429I12Rik; 3300002K07Rik; 9130020E21Rik; AI463719; C330049O21Rik; casein kinase 1 gamma 1; casein kinase 1 gamma 2; casein kinase 1 gamma 2 isoform; casein kinase 1 gamma 3; casein kinase 1 isoform gamma-2; casein kinase 1, gamma 1; casein kinase 1, gamma 2; casein kinase 1, gamma 3; casein kinase I isoform gamma-1; Casein kinase I isoform gamma-2; casein kinase I isoform gamma-3; casein kinase I, gamma 1; CK I gamma1; CK1G1; CK1G2; CK1gamma1; CKI gamma 1; CKI gamma1; CK-I gamma1; CKI- gamma1; CKI gamma-1; CKI-gamma 1; CKI-gamma 2; CKI-gamma 3; CKI-gamma-1; Csnk1g1; Csnk1g2; Csnk1g3; CSNK1G3L; KC1G1
Gene Symbols Csnk1g1, CSNK1G2, Csnk1g3
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 333-361 amino acids from the C-terminal region of mouse Csnk1g3.
Purification Method Antigen affinity chromatography, Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 103236, 214897, 70425
Target Species Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.