Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CK12 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501986113
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-6113 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-6113 Supplier Bioss Supplier No. BS4625R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

KRT12 encodes the type I intermediate filament chain keratin 12, expressed in corneal epithelia. Mutations in this gene lead to Meesmann corneal dystrophy.

Specifications

Antigen CK12
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene KRT12
Gene Accession No. Q99456
Gene Alias AI835216; CK-12; Cytokeratin-12; K12; Ka12; keratin 12; keratin 12 (Meesmann corneal dystrophy); keratin 12, type I; keratin complex 1, acidic, gene 12; keratin K12; Keratin, type I cytoskeletal 12; Keratin, type I cytoskeletal 12 (Cytokeratin 12); Keratin-12; Krt1-12; Krt12; Krt-12; krt12 {ECO:0000250; Type I keratin Ka12; UniProtKB:Q99456}
Gene Symbols KRT12
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human CK12/Cytokeratin 12.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3859
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.