Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CK18 Monoclonal Antibody (10H2), Bioss

Mouse Monoclonal Antibody
Supplier: Bioss BSM33103M

This item is not returnable.
View return policy
Description
CK18 Monoclonal antibody specifically detects CK18 in Human, Mouse samples. It is validated for Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western BlotSpecifications
CK18 | |
Monoclonal | |
1 mg/mL | |
TBS with 1% BSA, 50% glycerol and 0.03% ProClin 300 | |
P05783, P05784 | |
Mouse | |
Protein G | |
RUO | |
16668, 3875 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot | |
10H2 | |
Unconjugated | |
KRT18 | |
cb83; Cell proliferation-inducing gene 46 protein; CK 18; CK18; CK-18; CYK18; cytokeratin 18; cytokeratin endo B; cytokeratin type -1; DAPK-1; Cytokeratin18; cytokeratin-18; dapk1; dorsal aorta proneprin kinesin-1; DreK18; Endo B; I79_023185; K 18; K18; keratin 18; keratin 18, type I; keratin 18a, tandem duplicate 1; keratin complex 1, acidic, gene 18; keratin D; Keratin, type I cytoskeletal 18; Keratin18; keratin-18; Kerd; KRT 18; Krt1-18; KRT18; krt18 {ECO:0000250; krt18a.1; LOC100008885; PIG46; sb:cb83; type I cytoskeletal 18; UniProtKB:P05783}; wu:fa13f02; wu:fb36b04 | |
KLH conjugated synthetic peptide derived from human CK18 | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction