Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CK18 Monoclonal Antibody (10H2), Cyanine3, Bioss
SDP

Catalog No. 502281419
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-228-1419 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 50-228-1419 Supplier Bioss Supplier No. BSM33103MCY3
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

CK18 Monoclonal antibody specifically detects CK18 in Human, Mouse samples. It is validated for Immunocytochemistry

Specifications

Antigen CK18
Applications Immunocytochemistry
Classification Monoclonal
Clone 10H2
Concentration 1 mg/mL
Conjugate Cyanine3
Formulation TBS with 1% BSA, 50% glycerol and 0.03% ProClin 300
Gene KRT18
Gene Accession No. P05783, P05784
Gene Alias cb83; Cell proliferation-inducing gene 46 protein; CK 18; CK18; CK-18; CYK18; cytokeratin 18; cytokeratin endo B; cytokeratin type -1; DAPK-1; Cytokeratin18; cytokeratin-18; dapk1; dorsal aorta proneprin kinesin-1; DreK18; Endo B; I79_023185; K 18; K18; keratin 18; keratin 18, type I; keratin 18a, tandem duplicate 1; keratin complex 1, acidic, gene 18; keratin D; Keratin, type I cytoskeletal 18; Keratin18; keratin-18; Kerd; KRT 18; Krt1-18; KRT18; krt18 {ECO:0000250; krt18a.1; LOC100008885; PIG46; sb:cb83; type I cytoskeletal 18; UniProtKB:P05783}; wu:fa13f02; wu:fb36b04
Host Species Mouse
Immunogen KLH conjugated synthetic peptide derived from human CK18
Purification Method Protein G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 16668, 3875
Target Species Human, Mouse
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.