Learn More
Promega CK2 alpha 1 Kinase Enzyme System, 1mg
Catalog No. PRV6287 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
1 mg
Catalog No. Quantity
PRV6287 1 mg
1 options
Catalog No. PRV6287 Supplier Promega Supplier No. V6287
Add to Cart
Add to Cart
Request a Quote
Recombinant full-length human CK2 alpha 1 was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. CK2 alpha 1 is a serine-threonine protein kinase whose targets include many critical regulators of cellular growth.
  • Easily Screen and Profile CK2α1 Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP−Glo™ Assay for bioluminescent detection of kinase activity
Specifications
Quantity 1 mg
Content And Storage <-65°C