Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CK2 alpha-1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA587174
Description
Purity is > 95% by SDS-PAGE.
CSNK2A1 is a serine/threonine protein kinase that phosphorylates acidic proteins. The kinase exists as a tetramer and is composed of an alpha, an alpha-prime, and two beta subunits. The alpha subunits contain the catalytic activity while the beta subunits undergo autophosphorylation. The protein encoded by CK2 alpha-1 represents the alpha subunit. While CK2 alpha-1 is found on chromosome 20, a related transcribed pseudogene is found on chromosome 11. Three transcript variants encoding two different proteins have been found for CK2 alpha-1.
Specifications
CK2 alpha-1 | |
Polyclonal | |
Unconjugated | |
CSNK2A1 | |
casein kinase 2 alpha; casein kinase 2 alpha'; casein kinase 2 alpha 1; casein kinase 2 alpha 2; casein kinase 2, alpha 1 polypeptide; casein kinase 2, alpha prime polypeptide; casein kinase II alpha 1 polypeptide pseudogene; casein kinase II alpha 1 subunit; casein kinase II subunit alpha; Casein kinase II subunit alpha'; casein kinase II, alpha 1 polypeptide; casein kinase II, alpha 1 related sequence 4; CK II alpha; CK II alpha'; CK II alpha 3; CK2 catalytic subunit alpha; CK2A1; CK2A2; CK2alpha'; CKII; Ckiia; CSNK2A1; Csnk2a1-rs4; CSNK2A2; CSNK2A3; protein kinase CK2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
116549, 12995, 1457 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry, Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
P19139, P68400, Q60737 | |
CSNK2A1 | |
Recombinant protein of human CSNK2A1. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction