Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CK2 alpha-1 Recombinant Rabbit Monoclonal Antibody (2B6)
GREENER_CHOICE

Catalog No. FEMA549942
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEMA549942 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEMA549942 Supplier Invitrogen™ Supplier No. MA549942
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

CK2 alpha-1 Recombinant Monoclonal Antibody for Western Blot, ELISA

CSNK2A1 is a serine/threonine protein kinase that phosphorylates acidic proteins. The kinase exists as a tetramer and is composed of an alpha, an alpha-prime, and two beta subunits. The alpha subunits contain the catalytic activity while the beta subunits undergo autophosphorylation. The protein encoded by CK2 alpha-1 represents the alpha subunit. While CK2 alpha-1 is found on chromosome 20, a related transcribed pseudogene is found on chromosome 11. Three transcript variants encoding two different proteins have been found for CK2 alpha-1.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CK2 alpha-1
Applications ELISA, Western Blot
Classification Recombinant Monoclonal
Clone 2B6
Concentration 0.8 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene CSNK2A1
Gene Accession No. P68400, Q60737
Gene Alias casein kinase 2 alpha; casein kinase 2 alpha'; casein kinase 2 alpha 1; casein kinase 2 alpha 2; casein kinase 2, alpha 1 polypeptide; casein kinase 2, alpha prime polypeptide; casein kinase II alpha 1 polypeptide pseudogene; casein kinase II alpha 1 subunit; casein kinase II subunit alpha; Casein kinase II subunit alpha'; casein kinase II, alpha 1 polypeptide; casein kinase II, alpha 1 related sequence 4; CK II alpha; CK II alpha'; CK II alpha 3; CK2 catalytic subunit alpha; CK2A1; CK2A2; CK2alpha'; CKII; Ckiia; CSNK2A1; Csnk2a1-rs4; CSNK2A2; CSNK2A3; protein kinase CK2
Gene Symbols CSNK2A1
Host Species Rabbit
Immunogen A synthesized peptide derived from human CKII alpha.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 12995, 1457
Target Species Human, Mouse
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.