Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CKAP1 Antibody (1F6), mFluor Violet 610 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP231207MFV610
Description
CKAP1 Monoclonal antibody specifically detects CKAP1 in Human samples. It is validated for Flow CytometrySpecifications
CKAP1 | |
Monoclonal | |
mFluor Violet 610 SE | |
50mM Sodium Borate | |
Mouse | |
Protein A purified | |
RUO | |
1155 | |
Store at 4C in the dark. | |
IgG2b κ |
Flow Cytometry | |
1F6 | |
Flow Cytometry | |
CG22Tubulin-specific chaperone B, CKAP1MGC14625, CKAPI, cytoskeleton associated protein 1, Cytoskeleton-associated protein 1Cytoskeleton-associated protein CKAPI, tubulin folding cofactor B, tubulin-folding cofactor B | |
Recombinant human CKAP1 (1-244aa) purified from E. coli | |
0.1 mL | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction