Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CKAP1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP215907
Description
CKAP1 Polyclonal specifically detects CKAP1 in Human samples. It is validated for Western Blot.Specifications
CKAP1 | |
Polyclonal | |
Unconjugated | |
0.1M Tris (pH 7.0), 0.1M Glycine and 20% Glycerol with 0.01% Thimerosal | |
CG22Tubulin-specific chaperone B, CKAP1MGC14625, CKAPI, cytoskeleton associated protein 1, Cytoskeleton-associated protein 1Cytoskeleton-associated protein CKAPI, tubulin folding cofactor B, tubulin-folding cofactor B | |
Rabbit | |
27 kDa | |
0.1 mL | |
Primary | |
Human. Expected to cross react based on sequence identity: Mouse (87%), Rat (87%), Bovine (85%). | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Western Blot | |
0.71mg/mL | |
Western Blot 1:500-1:3000 | |
Q99426 | |
TBCB | |
Recombinant protein encompassing a sequence within the center region of human CKAP1. The exact sequence is proprietary. | |
Affinity Purified | |
RUO | |
1155 | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction