Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CKAP1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA531067
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA531067 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA531067 Supplier Invitrogen™ Supplier No. PA531067
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: NT2D1, U87-MG. Predicted reactivity: Mouse (87%), Rat (87%), Pig (87%), Bovine (85%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

CKAP1 binds to alpha-tubulin folding intermediates after their interaction with cytosolic chaperonin in the pathway leading from newly synthesized tubulin to properly folded heterodimer. Involved in regulation of tubulin heterodimer dissociation. May function as a negative regulator of axonal growth.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CKAP1
Applications Western Blot
Classification Polyclonal
Concentration 0.71 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7
Gene TBCB
Gene Accession No. Q99426
Gene Alias 2410007D12Rik; AU041393; CG22; Ckap1; CKAPI; cytoskeleton associated protein 1; cytoskeleton-associated protein 1; Cytoskeleton-associated protein CKAPI; liver cancer related protein; TBCB; tubulin folding cofactor B; tubulin-folding cofactor B; Tubulin-specific chaperone B; ZH14
Gene Symbols TBCB
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human CKAP1. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1155
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.