Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CKAP1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5118302
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5118302 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5118302 Supplier Invitrogen™ Supplier No. PA5118302
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

CKAP1 Polyclonal Antibody for Western Blot, ICC/IF, IP

CKAP1 binds to alpha-tubulin folding intermediates after their interaction with cytosolic chaperonin in the pathway leading from newly synthesized tubulin to properly folded heterodimer. Involved in regulation of tubulin heterodimer dissociation. May function as a negative regulator of axonal growth.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CKAP1
Applications Immunoprecipitation, Western Blot, Immunocytochemistry
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with 0.03% ProClin 300; pH 7
Gene TBCB
Gene Accession No. Q99426
Gene Alias 2410007D12Rik; AU041393; CG22; Ckap1; CKAPI; cytoskeleton associated protein 1; cytoskeleton-associated protein 1; Cytoskeleton-associated protein CKAPI; liver cancer related protein; TBCB; tubulin folding cofactor B; tubulin-folding cofactor B; Tubulin-specific chaperone B; ZH14
Gene Symbols TBCB
Host Species Rabbit
Immunogen E. coli-derived Human CKAP1/TBCB fragment.
Purification Method Antigen affinity chromatography, Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1155
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.