Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CKAP2 Antibody, Biotin, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP121391B
Description
CKAP2 Polyclonal antibody specifically detects CKAP2 in Human samples. It is validated for Western BlotSpecifications
CKAP2 | |
Polyclonal | |
Western Blot | |
CTCL tumor antigen se20-10, cytoskeleton associated protein 2, cytoskeleton-associated protein 2, LB1DKFZp686L1238, se20-10, TMAPFLJ10749, Tumor- and microtubule-associated protein, tumor-associated microtubule-associated protein | |
The immunogen recognized by this antibody maps to a region between residue 633 and 683 of human cytoskeleton associated protein 2 using the numbering given in entry NP_001091995.1 (GeneID 26586). | |
0.1 mL | |
Core ESC Like Genes, Stem Cell Markers | |
26586 | |
Store at 4°C in the dark. | |
IgG |
Western Blot | |
Biotin | |
PBS | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction