Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CKAP2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA552556
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA552556 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA552556 Supplier Invitrogen™ Supplier No. PA552556
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PPIRSHHSNT RDTVKQGISR TSANVTIRKG PHEKELLQSK TALSSVKTSS SQGIIRNKTL SRSIASEVVA RPASLSNDKL MEKSEPVDQR RHTAGKAIVD SRSAQPKETS EERKARLSEW KAGKGR Highest antigen sequence identity to the following orthologs: Mouse - 51%, Rat - 49%.

Nedd4-binding partner-1 (N4BP1) is a developmentally expressed protein interactor and monoubiquitylation substrate of Nedd4, a HECT domain-containing ubiquitin ligase that mediates ubiquitylation and proteasome degradation of target proteins and may have a fundamental role to play in embryonic processes. N4BP1 can undergo Nedd4-mediated polyubiquitylation and proteosomal degradation. N4BP1 also functions as a negative regulator of AIF4 (also known as ITCH or Atrophin-1), an E3-ubiquitin ligase, by interacting with the second WW domain of AIF4 and impairing ubiquitination of substrates.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CKAP2
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene CKAP2
Gene Accession No. Q8WWK9
Gene Alias AI461656; AW228814; Ckap2; CTCL tumor antigen se20-10; cytoskeleton associated protein 2; cytoskeleton-associated protein 2; LB1; se20-10; TMAP; Tumor- and microtubule-associated protein; tumor-associated microtubule-associated protein
Gene Symbols CKAP2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human CKAP2. Recombinant protein control fragment (Product #RP-89675).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26586
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.