Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CKAP2L Antibody, Biotin, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP121393B
Description
CKAP2L Polyclonal antibody specifically detects CKAP2L in Human samples. It is validated for Western BlotSpecifications
CKAP2L | |
Polyclonal | |
Western Blot | |
cytoskeleton associated protein 2-like, cytoskeleton-associated protein 2-like, FLJ40629, MGC39683 | |
The immunogen recognized by this antibody maps to a region between residue 200 and 250 of human cytoskeleton associated protein 2-like using the numbering given in entry NP_689728.3 (GeneID 150468). | |
0.1 mL | |
Primary | |
Human | |
Purified |
Western Blot | |
Biotin | |
PBS | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
150468 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction