Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CKAP2L Polyclonal Antibody, Invitrogen™

Catalog No. PIPA558778
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA558778 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA558778 Supplier Thermo Scientific Supplier No. PA558778

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Rabbit Polyclonal Antibody

Immunogen sequence: SSQVCIPQTS CVLQKSKAVS QRPNLTVGRF NSAIPSTPSI RPNGTSGNKH NNNGFQQKTQ TLDSKLKKAV PQNHFLNKTA PKTQADVTTV NGTQTN Highest antigen sequence identity to the following orthologs: Mouse - 59%, Rat - 57%.

The protein encoded by this gene is thought to be a mitotic spindle protein important to neural stem or progenitor cells. Mutations in this gene have been associated with spindle organization defects, including mitotic spindle defects, lagging chromosomes, and chromatin bridges. There is evidence that mutations in this gene are associated with Filippi syndrome, characterized by growth defects, microcephaly, intellectual disability, facial feature defects, and syndactyly. There is a pseudogene of this gene on chromosome 20. Alternative splicing results in multiple transcript variants.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CKAP2L
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene CKAP2L
Gene Accession No. Q8IYA6
Gene Alias CKAP2L; cytoskeleton associated protein 2 like; cytoskeleton associated protein 2-like; cytoskeleton-associated protein 2-like; FLJ40629; Radial fiber and mitotic spindle protein; radmis
Gene Symbols CKAP2L
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human CKAP2L
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 150468
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov

Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.