Learn More
CKAP2L Rabbit anti-Human, Polyclonal, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A302153AT

Description
The recommended shelf life for this product is 1 year from date of receipt.
The protein encoded by this gene is thought to be a mitotic spindle protein important to neural stem or progenitor cells. Mutations in this gene have been associated with spindle organization defects, including mitotic spindle defects, lagging chromosomes, and chromatin bridges. There is evidence that mutations in this gene are associated with Filippi syndrome, characterized by growth defects, microcephaly, intellectual disability, facial feature defects, and syndactyly. There is a pseudogene of this gene on chromosome 20. Alternative splicing results in multiple transcript variants.Specifications
CKAP2L | |
Polyclonal | |
Unconjugated | |
CKAP2L | |
cytoskeleton associated protein 2-like, FLJ40629, radial fiber and mitotic spindle, radial fiber and mitotic spindle protein, radmis | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
150468 | |
4° C | |
Liquid |
Immunoprecipitation | |
1 mg/ml | |
phosphate, tris citrate with 0.09% sodium azide; pH 7-8 | |
Q8IYA6 | |
CKAP2L | |
Between 200 and 250 | |
10 μL | |
Primary | |
Human | |
Antibody | |
IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.