Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CKAP4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA597591
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA597591 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA597591 Supplier Invitrogen™ Supplier No. PA597591
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

CKAP4 Polyclonal Antibody for Western Blot, ICC/IF, ELISA

This gene encodes the coiled-coil containing protein optineurin. Optineurin may play a role in normal-tension glaucoma and adult-onset primary open angle glaucoma. Optineurin interacts with adenovirus E3-14. 7K protein and may utilize tumor necrosis factor-alpha or Fas-ligand pathways to mediate apoptosis, inflammation or vasoconstriction. Optineurin may also function in cellular morphogenesis and membrane trafficking, vesicle trafficking, and transcription activation through its interactions with the RAB8, huntingtin, and transcription factor IIIA proteins. Alternative splicing results in multiple transcript variants encoding the same protein.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CKAP4
Applications ELISA, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 4.085 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene CKAP4
Gene Accession No. Q07065
Gene Alias 5630400A09Rik; 63 kDa membrane protein; 63-kDa cytoskeleton-linking membrane protein; CKAP4; Climp-63; cytoskeleton associated protein 4; cytoskeleton-associated protein 4; ERGIC-63; LPC-1; p63; transmembrane protein (63kD), endoplasmic reticulum/Golgi intermediate compartment; type-II transmembrane protein p63
Gene Symbols CKAP4
Host Species Rabbit
Immunogen Recombinant Human Cytoskeleton-associated protein 4 protein (260-394AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10970
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.