Learn More
CKAP4 Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A302257AT

Description
The recommended shelf life for this product is 1 year from date of receipt. Application Note: For IHC, epitope retrieval with citrate buffer pH 6.0 is recommended for FFPE tissue sections.
CKAP4/p63 is a substrate of DHHC2, a putative tumor suppressor. CK MNF116 and p63 were useful in identifying squamous cell carcinomas with single cell infiltration. Those results suggested a role of CKAP4/p63 in tumor.Specifications
CKAP4 | |
Polyclonal | |
TBS with 0.1% BSA and 0.09% sodium azide | |
Q07065, Q8BMK4 | |
Ckap4 | |
Between 500 and 550 | |
20 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Immunohistochemistry, Western Blot | |
Unconjugated | |
CKAP4 | |
CLIMP-63, ERGIC-63, p63, 63 kDa membrane protein, 63-kDa cytoskeleton-linking membrane protein, transmembrane protein (63kD), endoplasmic reticulum/Golgi intermediate compartment, type-II transmembrane protein p63 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
10970, 216197 | |
4° C | |
Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.