Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CKAP5 Monoclonal Antibody (3C13)
GREENER_CHOICE

Catalog No. PIMA549287
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA549287 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA549287 Supplier Invitrogen™ Supplier No. MA549287
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Adding 0.2 mL of distilled water will yield a concentration of 500 μg/mL. Positive Control - WB: human U-87MG whole cell, human COLO-320 whole cell, human SW620 whole cell, human HEPG2 whole cell, human CACO-2 whole cell, rat brain tissue, mouse brain tissue. IHC: human liver cancer tissue, human rectal cancer tissue, human renal cancer tissue, rat brain tissue. ICC/IF: Hela cell. Flow: A549 cell. Store at -20°C for one year from date of receipt. After reconstitution, at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for six months. Avoid repeated freeze-thaw cycles.

Colonic and hepatic tumor over-expressed protein (ch-TOG) belongs to the TOG/XMAP215/Dis1 evolutionarily ancient family of microtubule-associated proteins with isoforms found in all major kingdoms of eukaryotes. This protein is required for the assembly and function of the meiotic or mitotic spindles during cell division and is localized to microtubule-organizing centers (MTOCs) during interphase or to the spindle poles and centrosomes during mitosis or cytokinesis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CKAP5
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 3C13
Concentration 500 μg/mL
Conjugate Unconjugated
Formulation PBS with 4mg trehalose and no preservative
Gene CKAP5
Gene Accession No. A2AGT5, Q14008
Gene Alias 3110043H24Rik; 4930432B04Rik; CHTOG; ch-TOG; ch-TOGp; CKAP5; colonic and hepatic tumor overexpressed gene protein; colonic and hepatic tumor over-expressed gene protein; colonic and hepatic tumor over-expressed protein; cytoskeleton associated protein 5; cytoskeleton-associated protein 5; D730027C18Rik; KIAA0097; mKIAA0097; MSPS; T25636; TOG; TOGp
Gene Symbols CKAP5
Host Species Mouse
Immunogen E.coli-derived human ch TOG/CKAP5 recombinant protein (Position: M1-L221).
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 311191, 75786, 9793
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Lyophilized
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.