Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CKAP5 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA559150
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA559150 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA559150 Supplier Invitrogen™ Supplier No. PA559150
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MSSKLNQARS MSGHPEAAQM VRREFQLDLD EIENDNGTVR CEMPELVQHK LDDIFEPVLI PEPKIRAVSP HFDDMHSNTA STINFI Highest antigen sequence identity to the following orthologs: Mouse - 99%, Rat - 100%.

Colonic and hepatic tumor over-expressed protein (ch-TOG) belongs to the TOG/XMAP215/Dis1 evolutionarily ancient family of microtubule-associated proteins with isoforms found in all major kingdoms of eukaryotes. This protein is required for the assembly and function of the meiotic or mitotic spindles during cell division and is localized to microtubule-organizing centers (MTOCs) during interphase or to the spindle poles and centrosomes during mitosis or cytokinesis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CKAP5
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene CKAP5
Gene Accession No. Q14008
Gene Alias 3110043H24Rik; 4930432B04Rik; CHTOG; ch-TOG; ch-TOGp; CKAP5; colonic and hepatic tumor overexpressed gene protein; colonic and hepatic tumor over-expressed gene protein; colonic and hepatic tumor over-expressed protein; cytoskeleton associated protein 5; cytoskeleton-associated protein 5; D730027C18Rik; KIAA0097; mKIAA0097; MSPS; T25636; TOG; TOGp
Gene Symbols CKAP5
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human CKAP5. Recombinant protein control fragment (Product #RP-97496).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9793
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.