Learn More
Invitrogen™ CKAP5 Polyclonal Antibody
Description
PA1-32456 detects CKAP5 from rabbit and mammal samples.
Specifications
Specifications
| Antigen | CKAP5 |
| Applications | Immunoprecipitation, Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | Conc. Not Determined |
| Conjugate | Unconjugated |
| Formulation | Whole serum with no preservative |
| Gene | CKAP5 |
| Gene Accession No. | A2AGT5, Q14008 |
| Gene Alias | 3110043H24Rik; 4930432B04Rik; CHTOG; ch-TOG; ch-TOGp; CKAP5; colonic and hepatic tumor overexpressed gene protein; colonic and hepatic tumor over-expressed gene protein; colonic and hepatic tumor over-expressed protein; cytoskeleton associated protein 5; cytoskeleton-associated protein 5; D730027C18Rik; KIAA0097; mKIAA0097; MSPS; T25636; TOG; TOGp |
| Gene Symbols | CKAP5 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.